Tributyl phosphite 94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tri-n-butylphosphite
L14307-AE
L14307-AP
CAAAAL14307-AEEA
75.52
CAD
CAAAAL14307-AE
CAAAAL14307-AP
Tributyl phosphite 94%
Tributyl phosphite
Purity: 94%
CAS No.: 102-85-2
Molecular Formula: C12H27O3P
Formula Weight: 250.32
Flash Point: 82°(179°F)
Refractive Index: 1.4320
Melting Point: -80°
Boiling Point: 125-128°/10mm
Beilstein Reference: 1703866
MDL No.: MFCD00009437
Formula:
C₁₂H₂₇O₃P Boiling Pt: 125…128 °C/10 mm Melting Pt: –80 °C Density: 0.915 Flash Pt: 82 °C(179 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009437 CAS Number: 102-85-2 EINECS: 203-061-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...