4-(Trifluoromethyl)benzhydrol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Phenyl(4-(trifluoromethyl)phenyl)methanol
L14147-03
L14147-06
CAAAAL14147-03EA
27.49
CAD
CAAAAL14147-03
CAAAAL14147-06
4-(Trifluoromethyl)benzhydrol 97%
4-(Trifluoromethyl)benzhydrol
Purity: 97%
CAS No.: 395-23-3
Molecular Formula: C14H11F3O
Formula Weight: 252.23
Melting Point: 59-61°
Boiling Point: 122-124°/0.4mm
MDL No.: MFCD00014399
Formula:
C₁₄H₁₁F₃O MW: 252.23 g/mol Boiling Pt: 122…124 °C/0.4 mm Melting Pt: 59…61 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014399 CAS Number: 395-23-3 EINECS: 209-917-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...