7-Ethoxycoumarin 99%

Supplier: Thermo Scientific Chemicals

L14116-MC L14116-ME
CAAAAL14116-MCEA 83.74 CAD
CAAAAL14116-MC CAAAAL14116-ME
7-Ethoxycoumarin 99%
7-Ethoxycoumarin

Purity: 99% CAS No.: 31005-02-4 Molecular Formula: C11H10O3 Formula Weight: 190.20 Melting Point: 88-91° Beilstein Reference: 146200

Formula: C₁₁H₁₀O₃
Melting Pt: 88…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00006877
CAS Number: 31005-02-4
EINECS: 250-429-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR