3-Aminocoumarin 97%

Supplier: Thermo Scientific Chemicals
Warning

L14031-04 L14031-MC L14031-ME
CAAAAL14031-04EA 417.12 CAD
CAAAAL14031-04 CAAAAL14031-MC CAAAAL14031-ME
3-Aminocoumarin 97%
3-Aminocoumarin

Purity: 97% CAS No.: 1635-31-0 Molecular Formula: C9H7NO2 Formula Weight: 161.16 Melting Point: 135-137° Beilstein Reference: 128033

Formula: C₉H₇NO₂
Melting Pt: 135…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00016965
CAS Number: 1635-31-0
EINECS: 216-659-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR