2-Acetylbenzofuran 99%

Supplier: Thermo Scientific Chemicals

L13771-06 L13771-14
CAAAAL13771-06EA 95.27 CAD
CAAAAL13771-06 CAAAAL13771-14
2-Acetylbenzofuran 99%
2-Acetylbenzofuran

Purity: 99% CAS No.: 1646-26-0 Molecular Formula: C10H8O2 Formula Weight: 160.17 Flash Point: 110°(230°F) Melting Point: 70-72° Boiling Point: 110-113°/3mm Beilstein Reference: 117910 MDL No.: MFCD00005849

Formula: C₁₀H₈O₂
Boiling Pt: 110…113 °C/3 mm
Melting Pt: 70…72 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005849
CAS Number: 1646-26-0
EINECS: 216-706-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR