Pyrrole-2-carboxylic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L13756-03
L13756-06
L13756-14
CAAAAL13756-03EA
68.57
CAD
CAAAAL13756-03
CAAAAL13756-06
CAAAAL13756-14
Pyrrole-2-carboxylic acid 99%
Pyrrole-2-carboxylic acid
Purity: 99%
CAS No.: 634-97-9
Molecular Formula: C5H5NO2
Formula Weight: 111.10
Melting Point: ca 205° subl.
Beilstein Reference: 80825
Formula:
C₅H₅NO₂ Melting Pt: ca 205 °C (subl.) Storage Temperature: Ambient |
MDL Number:
MFCD00005219 CAS Number: 634-97-9 EINECS: 211-221-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...