N-Ethylpyridine-4-methylamine 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-(4-Pyridylmethyl)ethylamine, 4-(Ethylaminomethyl)pyridine

L13714-06 L13714-14
CAAAAL13714-06EA 49.14 CAD
CAAAAL13714-06 CAAAAL13714-14
N-Ethylpyridine-4-methylamine 96%
N-Ethylpyridine-4-methylamine

Purity: 96% CAS No.: 33403-97-3 Molecular Formula: C8H12N2 Formula Weight: 136.20 Flash Point: 89°(192°F) Refractive Index: 1.5175 Boiling Point: 103-105°/8mm Beilstein Reference: 471685 MDL No.: MFCD00023632

Formula: C₈H₁₂N₂
MW: 136.20 g/mol
Boiling Pt: 103…105 °C (8 mmHg)
Density: 0.98
Flash Pt: 89 °C (192 °F)
Storage Temperature: Ambient
MDL Number: MFCD00023632
CAS Number: 33403-97-3
EINECS: 251-506-5
UN: 2735
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR