2-(4-tert-Butylphenyl)ethanol 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L13611-06
L13611-14
CAAAAL13611-06EA
95.27
CAD
CAAAAL13611-06
CAAAAL13611-14
2-(4-tert-Butylphenyl)ethanol 96%
2-(4-tert-Butylphenyl)ethanol
Purity: 96%
CAS No.: 5406-86-0
Molecular Formula: C12H18O
Formula Weight: 178.28
Refractive Index: 1.5209
Melting Point: 31-35°
Boiling Point: 141-143°/15mm
Beilstein Reference: 2246595
UN No.: 3077
MDL No.: MFCD00236023
Formula:
C₁₂H₁₈O Boiling Pt: 141…143 °C/15 mm Melting Pt: 31…35 °C Density: 0.975 Storage Temperature: Ambient |
MDL Number:
MFCD00236023 CAS Number: 5406-86-0 EINECS: 410-020-5 UN: 3077 ADR: "9",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...