Diethyl decylphosphonate 97%
Supplier: Thermo Scientific Chemicals
L13038-06
L13038-14
CAAAAL13038-06EA
102.7
CAD
CAAAAL13038-06
CAAAAL13038-14
Diethyl decylphosphonate 97%
Diethyl decylphosphonate
Purity: 97%
CAS No.: 16165-68-7
Molecular Formula: C14H31O3P
Formula Weight: 278.38
Refractive Index: 1.4390
Boiling Point: 128-130°/0.2mm
MDL No.: MFCD00015593
Formula:
C₁₄H₃₁O₃P Boiling Pt: 128…130 °C/0.2 mm Storage Temperature: Ambient |
MDL Number:
MFCD00015593 CAS Number: 16165-68-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...