7-Phenyl-1-heptanol 97%
Supplier: Thermo Scientific Chemicals
L12533-03
L12533-06
CAAAAL12533-03EA
136.99
CAD
CAAAAL12533-03
CAAAAL12533-06
7-Phenyl-1-heptanol 97%
7-Phenyl-1-heptanol
Purity: 97%
CAS No.: 3208-25-1
Molecular Formula: C6H5(CH2)7OH
Formula Weight: 192.30
Refractive Index: 1.5080
Boiling Point: 115-117°/0.5mm
MDL No.: MFCD00037315
Formula:
C₁₃H₂₀O Boiling Pt: 115…117 °C (0,5 mmHg) Density: 0,962 Storage Temperature: Ambient |
MDL Number:
MFCD00037315 CAS Number: 3208-25-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...