Phthalamic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

L12432-06 L12432-14
CAAAAL12432-06EA 70.47 CAD
CAAAAL12432-06 CAAAAL12432-14
Phthalamic acid 99%
Phthalamic acid

Purity: 99% CAS No.: 88-97-1 Molecular Formula: C8H7NO3 Formula Weight: 165.15 Melting Point: 140-143° Beilstein Reference: 1868205

Formula: C₈H₇NO₃
Melting Pt: 140…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00025476
CAS Number: 88-97-1
EINECS: 201-871-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR