(4-Cyanobenzyl)triphenylphosphonium chloride 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L12187-06
L12187-14
CAAAAL12187-06EA
60.36
CAD
CAAAAL12187-06
CAAAAL12187-14
(4-Cyanobenzyl)triphenylphosphonium chloride 99%
(4-Cyanobenzyl)triphenylphosphonium chloride
Purity: 99%
CAS No.: 20430-33-5
Molecular Formula: C26H21ClNP
Formula Weight: 413.89
Odor: Odorless
Melting Point: 322-326°
Beilstein Reference: 4630137
MDL No.: MFCD00011752
Formula:
C₂₆H₂₁ClNP Melting Pt: 322…326 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011752 CAS Number: 20430-33-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...