1-Benzhydrylpiperazine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L11530-14
L11530-22
CAAAAL11530-14EA
62.73
CAD
CAAAAL11530-14
CAAAAL11530-22
1-Benzhydrylpiperazine 97%
1-Benzhydrylpiperazine
Purity: 97%
CAS No.: 841-77-0
Molecular Formula: C17H20N2
Formula Weight: 252.36
Melting Point: 91-92°
Beilstein Reference: 222773
Formula:
C₁₇H₂₀N₂ Melting Pt: 91…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00038379 CAS Number: 841-77-0 EINECS: 212-667-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...