5-Acetamido-2-aminobenzoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L11256-06
L11256-14
CAAAAL11256-06EA
67.31
CAD
CAAAAL11256-06
CAAAAL11256-14
5-Acetamido-2-aminobenzoic acid 97%
5-Acetamido-2-aminobenzoic acid
Purity: 97%
CAS No.: 50670-83-2
Molecular Formula: C9H10N2O3
Formula Weight: 194.19
Melting Point: ca 230° dec.
Beilstein Reference: 2726882
Formula:
C₉H₁₀N₂O₃ MW: 194.19 g/mol Melting Pt: ca 230 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00060120 CAS Number: 50670-83-2 EINECS: 256-702-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...