Norbornene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Norbornylene
L11103-22
L11103-36
CAAAAL11103-22EA
63.2
CAD
CAAAAL11103-22
CAAAAL11103-36
Norbornene 99%
Norbornene
Purity: 99%
CAS No.: 498-66-8
Molecular Formula: C7H10
Formula Weight: 94.16
Flash Point: -15°(5°F)
Odor: Pungent
Boiling Point: 95-96°
Beilstein Reference: 1098544
UN No.: 1325
MDL No.: MFCD00082304
Formula:
C₇H₁₀ Boiling Pt: 95…96 °C Melting Pt: 44…47 °C Density: 0.955 Flash Pt: -15 °C(5 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00082304 CAS Number: 498-66-8 EINECS: 207-866-0 UN: 1325 ADR: "4.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...