Chloromethyl methyl sulfide 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L10858-09
L10858-18
CAAAAL10858-09EA
109.18
CAD
CAAAAL10858-09
CAAAAL10858-18
Chloromethyl methyl sulfide 95%
Chloromethyl methyl sulphide
Fieser: 4,84 6,109 8,94 9,107 12,117
Purity: 95%
CAS No.: 2373-51-5
Molecular Formula: CH3SCH2Cl
Formula Weight: 96.58
Storage Temperature: 2°C to 8°C
Flash Point: 17°(62°F)
Refractive Index: 1.4980
Odor: Stench
Boiling Point: 104-105°
Beilstein Reference: 1730851
UN No.: 1993
MDL No.: MFCD00000923
Formula:
C₂H₅ClS Boiling Pt: 104…105 °C Density: 1.160 Flash Pt: 17 °C(62 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000923 CAS Number: 2373-51-5 EINECS: 219-148-4 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...