4-Ethylphenylacetonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L10855-03
L10855-06
CAAAAL10855-03EA
79.63
CAD
CAAAAL10855-03
CAAAAL10855-06
4-Ethylphenylacetonitrile 97%
4-Ethylphenylacetonitrile
Purity: 97%
CAS No.: 51632-28-1
Molecular Formula: C10H11N
Formula Weight: 145.21
Refractive Index: 1.5164
Boiling Point: 110°/1mm
UN No.: 3276
MDL No.: MFCD00040893
Formula:
C₁₀H₁₁N Boiling Pt: 110 °C/1 mm Density: 0.970 Storage Temperature: Ambient |
MDL Number:
MFCD00040893 CAS Number: 51632-28-1 UN: 3276 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...