Triethyl-4-phosphonobutyrate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L10488-04
L10488-09
CAAAAL10488-04EA
60.36
CAD
CAAAAL10488-04
CAAAAL10488-09
Triethyl-4-phosphonobutyrate 97%
Triethyl 4-phosphonobutyrate
Purity: 97%
CAS No.: 2327-69-7
Molecular Formula: C10H21O5P
Formula Weight: 252.25
Refractive Index: 1.4370
Boiling Point: 117-119°/0.4mm
MDL No.: MFCD00015168
Formula:
C₁₀H₂₁O₅P Boiling Pt: 117…119 °C (0,4 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00015168 CAS Number: 2327-69-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...