Tantalum(V) ethanolate 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L10288-06
L10288-14
CAAAAL10288-06EA
183.28
CAD
CAAAAL10288-06
CAAAAL10288-14
Tantalum(V) ethanolate 99+%
Tantalum(V) ethanolate
Purity: 99+%
CAS No.: 6074-84-6
Molecular Formula: C10H25O5Ta
Formula Weight: 406.25
Flash Point: 30°(86°F)
Refractive Index: 1.4880
Melting Point: 18-19°
Boiling Point: 145°/0.1mm
Beilstein Reference: 3678999
UN No.: 2924
MDL No.: MFCD00049785
Formula:
C₁₀H₂₅O₅Ta Boiling Pt: 145 °C/0.1 mm Melting Pt: 18…19 °C Density: 1.566 Flash Pt: 30 °C(86 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00049785 CAS Number: 6074-84-6 EINECS: 228-010-2 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...