Cyclohexene-1-carbonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L10284-04
L10284-09
CAAAAL10284-04EA
96.85
CAD
CAAAAL10284-04
CAAAAL10284-09
Cyclohexene-1-carbonitrile 98%
Cyclohexene-1-carbonitrile
Purity: 98%
CAS No.: 1855-63-6
Molecular Formula: C7H9N
Formula Weight: 107.16
Flash Point: 83°(181°F)
Refractive Index: 1.4820
Boiling Point: 85-88°/17mm
Beilstein Reference: 385837
UN No.: 3276
MDL No.: MFCD00013772
Formula:
C₇H₉N Boiling Pt: 85…88 °C/17 mm Density: 0.947 Flash Pt: 83 °C(181 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013772 CAS Number: 1855-63-6 EINECS: 217-454-2 UN: 3276 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...