Triethyl-3-phosphonopropionate 98%

Supplier: Thermo Scientific Chemicals
Warning

L10190-09 L10190-18
CAAAAL10190-09EA 92.11 CAD
CAAAAL10190-09 CAAAAL10190-18
Triethyl-3-phosphonopropionate 98%
Triethyl 3-phosphonopropionate

Purity: 98% CAS No.: 3699-67-0 Molecular Formula: C9H19O5P Formula Weight: 238.22 Flash Point: 110°(230°F) Refractive Index: 1.4330 Boiling Point: 127-129°/2mm Beilstein Reference: 1790966 MDL No.: MFCD00015167

Formula: C₉H₁₉O₅P
Boiling Pt: 127…129 °C/2 mm
Density: 1.094
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015167
CAS Number: 3699-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR