3,5-Xylylacetonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

L10170-04 L10170-09
CAAAAL10170-04EA 115.97 CAD
CAAAAL10170-04 CAAAAL10170-09
3,5-Xylylacetonitrile 98%
3,5-Xylylacetonitrile

Purity: 98% CAS No.: 39101-54-7 Molecular Formula: C10H11N Formula Weight: 145.20 Melting Point: 42-45° Boiling Point: 170°/20mm UN No.: 3439 MDL No.: MFCD00060304

Formula: C₁₀H₁₁N
Boiling Pt: 170 °C/20 mm
Melting Pt: 42…45 °C
Storage Temperature: Ambient
MDL Number: MFCD00060304
CAS Number: 39101-54-7
EINECS: 254-292-1
UN: 3439
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR