2,5-Di-tert-butyl-p-benzoquinone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,5-Di-tert-butylcyclohexa-2,5-diene-1,4-dione

L10054-06 L10054-14
CAAAAL10054-06EA 65.73 CAD
CAAAAL10054-06 CAAAAL10054-14
2,5-Di-tert-butyl-p-benzoquinone 99%
2,5-Di-tert-butyl-p-benzoquinone

Purity: 99% CAS No.: 2460-77-7 Molecular Formula: C14H20O2 Formula Weight: 220.31 Melting Point: 152-153° Beilstein Reference: 2047945

Formula: C₁₄H₂₀O₂
MW: 220.31 g/mol
Melting Pt: 152…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00019442
CAS Number: 2460-77-7
EINECS: 219-552-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR