2-(Methylthio)nicotinic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09920-04
L09920-09
CAAAAL09920-04EA
22.12
CAD
CAAAAL09920-04
CAAAAL09920-09
2-(Methylthio)nicotinic acid 98+%
2-(Methylthio)nicotinic acid
Purity: 98+%
CAS No.: 74470-23-8
Molecular Formula: C7H7NO2S
Formula Weight: 169.20
Melting Point: 215-218°
Beilstein Reference: 4391503
Formula:
C₇H₇NO₂S MW: 169.20 g/mol Melting Pt: 215…218 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010101 CAS Number: 74470-23-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...