Dibenzyl phosphonate 90+%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09858-06
L09858-14
L09858-22
CAAAAL09858-06EA
57.04
CAD
CAAAAL09858-06
CAAAAL09858-14
CAAAAL09858-22
Dibenzyl phosphonate 90+%, Technical Grade
Dibenzyl phosphonate
Fieser: 1,198
Purity: 90+%
CAS No.: 17176-77-1
Molecular Formula: C14H15O3P
Formula Weight: 262.25
Flash Point:
Formula:
C₁₄H₁₅O₃P Boiling Pt: 110…120 °C/0.1 mm Density: 1.187 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004774 CAS Number: 17176-77-1 EINECS: 241-226-1 Merck Index: 14,3014 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...