(2-Chlorophenyl)acetone 96%
Supplier: Thermo Scientific Chemicals
L09846-03
L09846-06
CAAAAL09846-03EA
45.5
CAD
CAAAAL09846-03
CAAAAL09846-06
(2-Chlorophenyl)acetone 96%
(2-Chlorophenyl)acetone
Purity: 96%
CAS No.: 6305-95-9
Molecular Formula: C9H9ClO
Formula Weight: 168.62
Refractive Index: 1.5360
Boiling Point: 102-103°/3mm
MDL No.: MFCD00045175
Formula:
C₉H₉ClO Boiling Pt: 102…103 °C/3 mm Storage Temperature: Ambient |
MDL Number:
MFCD00045175 CAS Number: 6305-95-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...