FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3',6'-Dihydroxy-5-isothiocyanato-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 5-FITC, Fluorescein Isothiocyanate, 5-isothiocyanatofluorescein, Fluorescein Isothiocyanate, Isomer I
L09319-03
L09319-MD
L09319-MF
CAAAAL09319-03EA
492.96
CAD
CAAAAL09319-03
CAAAAL09319-MD
CAAAAL09319-MF
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) 95%
FITC (Fluorescein 5-isothiocyanate, Fluorescein isothiocyanate isomer I)
Useful for protein labelling.
Purity: 95%
CAS No.: 3326-32-7
Molecular Formula: C21H11NO5S
Formula Weight: 389.39
Storage Temperature: 2°C to 8°C
Melting Point:
Formula:
C₂₁H₁₁NO₅S Melting Pt: >360 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005063 CAS Number: 3326-32-7 EINECS: 222-042-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...