Methyl-2-methylnicotinate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09074-06
L09074-14
CAAAAL09074-06EA
137.93
CAD
CAAAAL09074-06
CAAAAL09074-14
Methyl-2-methylnicotinate 97%
Methyl 2-methylnicotinate
Purity: 97%
CAS No.: 65719-09-7
Molecular Formula: C8H9NO2
Formula Weight: 151.17
Refractive Index: 1.5165
Boiling Point: 102-104°/10mm
MDL No.: MFCD00051961
Formula:
C₈H₉NO₂ Boiling Pt: 102…104 °C/10 mm Storage Temperature: Ambient |
MDL Number:
MFCD00051961 CAS Number: 65719-09-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...