Phthaloyl dichloride 94%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L08753-22
L08753-36
CAAAAL08753-22EA
99.7
CAD
CAAAAL08753-22
CAAAAL08753-36
Phthaloyl dichloride 94%
Phthaloyl dichloride
Fieser: 1,882 14,263
Purity: 94%
CAS No.: 88-95-9
Molecular Formula: C8H4Cl2O2
Formula Weight: 203.03
Flash Point: 151°(304°F)
Refractive Index: 1.5680
Melting Point: 12°
Boiling Point: 276°
Beilstein Reference: 608200
Merck Reference: 14,7375
UN No.: 3265
MDL No.: MFCD00000666
Formula:
C₈H₄Cl₂O₂ Boiling Pt: 276 °C Melting Pt: 12 °C Density: 1.409 Flash Pt: 151 °C(304 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000666 CAS Number: 88-95-9 EINECS: 201-869-0 UN: 3265 ADR: "8",III Merck Index: 14,7375 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...