4-tert-Butyl-2,6-dinitrophenol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L08670-06
L08670-14
CAAAAL08670-06EA
27.49
CAD
CAAAAL08670-06
CAAAAL08670-14
4-tert-Butyl-2,6-dinitrophenol 97%
4-tert-Butyl-2,6-dinitrophenol
Purity: 97%
CAS No.: 4097-49-8
Molecular Formula: C10H12N2O5
Formula Weight: 240.22
Melting Point: 93-95°
Beilstein Reference: 1986466
UN No.: 2811
MDL No.: MFCD00051969
Formula:
C₁₀H₁₂N₂O₅ Melting Pt: 93…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051969 CAS Number: 4097-49-8 EINECS: 223-856-9 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...