Potassium 2,5-dihydroxybenzenesulfonate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Hydroquinonesulfonic acid potassium salt, Potassium 2,5-dihydroxybenzenesulfonate, 2,5-Dihydroxybenzenesulfonic acid potassium sal, 2,5-Dihydroxybenzenesulphonic acid potassium sal, Hydroquinonesulphonic acid potassium salt, Potassium 2,5-dihydroxybenzenesulphonate
L08406-22
L08406-36
CAAAAL08406-22EA
60.2
CAD
CAAAAL08406-22
CAAAAL08406-36
Potassium 2,5-dihydroxybenzenesulfonate 98+%
Potassium 2,5-dihydroxybenzenesulphonate
Purity: 98+%
CAS No.: 21799-87-1
Molecular Formula: C6H5KO5S
Formula Weight: 228.27
Melting Point: ca 251° dec.
Beilstein Reference: 5187027
Formula:
C₆H₅KO₅S Melting Pt: ca 251 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00007475 CAS Number: 21799-87-1 EINECS: 244-584-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...