1,2,3,4-Tetrahydroisoquinoline 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L08143-14
L08143-22
CAAAAL08143-14EA
77.26
CAD
CAAAAL08143-14
CAAAAL08143-22
1,2,3,4-Tetrahydroisoquinoline 97%
1,2,3,4-Tetrahydroisoquinoline
Purity: 97%
CAS No.: 91-21-4
Molecular Formula: C9H11N
Formula Weight: 133.19
Flash Point: 98°(208°F)
Refractive Index: 1.5680
Odor: Amine-like
pH: 11
Melting Point: -30°
Boiling Point: 232-233°
Beilstein Reference: 116156
MDL No.: MFCD00006896
Formula:
C₉H₁₁N MW: 133.19 g/mol Boiling Pt: 232…233 °C Melting Pt: –30 °C Density: 1.049 Flash Pt: 98 °C (208 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006896 CAS Number: 91-21-4 EINECS: 202-050-0 UN: 2735 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...