Methyl-2,6-difluorobenzoate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L08063-06
L08063-14
CAAAAL08063-06EA
61.46
CAD
CAAAAL08063-06
CAAAAL08063-14
Methyl-2,6-difluorobenzoate 97%
Methyl 2,6-difluorobenzoate
Purity: 97%
CAS No.: 13671-00-6
Molecular Formula: C8H6F2O2
Formula Weight: 172.13
Flash Point: 95°(203°F)
Refractive Index: 1.4760
Boiling Point: 203-204°
Beilstein Reference: 1948618
MDL No.: MFCD00051778
Formula:
C₈H₆F₂O₂ Boiling Pt: 203…204 °C Density: 1.281 Flash Pt: 95 °C(203 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051778 CAS Number: 13671-00-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...