4-Nitrophthalic anhydride tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Nitroisobenzofuran-1,3-dione

L08020-06 L08020-14
CAAAAL08020-06EA 100.17 CAD
CAAAAL08020-06 CAAAAL08020-14
4-Nitrophthalic anhydride tech. 90%, Technical Grade
4-Nitrophthalic anhydride

Fieser: 18,253


Purity: 90% CAS No.: 5466-84-2 Molecular Formula: C8H3NO5 Formula Weight: 193.11 Melting Point: 116-121° Beilstein Reference: 179682

Formula: C₈H₃NO₅
MW: 193.12 g/mol
Melting Pt: 116…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00005922
CAS Number: 5466-84-2
EINECS: 226-776-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR