3-Hydroxypicolinic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Hydroxypyridine-2-carboxylic acid
L07489-03
L07489-06
CAAAAL07489-03EA
75.84
CAD
CAAAAL07489-03
CAAAAL07489-06
3-Hydroxypicolinic acid 98%
3-Hydroxypicolinic acid
Purity: 98%
CAS No.: 874-24-8
Molecular Formula: C6H5NO3
Formula Weight: 139.11
Melting Point: ca 214° dec.
Beilstein Reference: 118954
Formula:
C₆H₅NO₃ MW: 139.11 g/mol Melting Pt: ∼ 214 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00006294 CAS Number: 874-24-8 EINECS: 212-859-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...