Coumarin-3-carboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Oxo-2H-chromene-3-carboxylic acid

L07133-14 L07133-22
CAAAAL07133-14EA 103.17 CAD
CAAAAL07133-14 CAAAAL07133-22
Coumarin-3-carboxylic acid 98%
Coumarin-3-carboxylic acid

Purity: 98% CAS No.: 531-81-7 Molecular Formula: C10H6O4 Formula Weight: 190.15 Melting Point: 190-193° Beilstein Reference: 154276 Merck Reference: 14,2563 UN No.: 2811 MDL No.: MFCD00006852

Formula: C₁₀H₆O₄
MW: 190.16 g/mol
Melting Pt: 190…193 °C
Storage Temperature: Ambient
MDL Number: MFCD00006852
CAS Number: 531-81-7
EINECS: 208-518-0
UN: 2811
ADR: 6.1,III
Merck Index: 14,2563

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR