ε-Caprolactam 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Azepan-2-one, 2-Oxohexamethyleneimine, 6-Aminocaproic acid lactam
L06999-0E
L06999-22
L06999-36
CAAAAL06999-0EEA
139.67
CAD
CAAAAL06999-0E
CAAAAL06999-22
CAAAAL06999-36
ε-Caprolactam 99%
ε-Caprolactam
Fieser: 9,316
Purity: 99%
CAS No.: 105-60-2
Molecular Formula: C6H11NO
Formula Weight: 113.16
Flash Point: 152°(305°F)
pH: 7-8.5
Melting Point: 68-72°
Boiling Point: 270°
Beilstein Reference: 106934
Merck Reference: 14,1761
MDL No.: MFCD00006936
Formula:
C₆H₁₁NO MW: 113.16 g/mol Boiling Pt: 270 °C Melting Pt: 68…72 °C Density: 1.01 Flash Pt: 152 °C(305 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006936 CAS Number: 105-60-2 EINECS: 203-313-2 Merck Index: 14,1761 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...