2-Furoyl chloride 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L06960-18
L06960-30
CAAAAL06960-18EA
110.76
CAD
CAAAAL06960-18
CAAAAL06960-30
2-Furoyl chloride 98+%
2-Furoyl chloride
Purity: 98+%
CAS No.: 527-69-5
Molecular Formula: C5H3ClO2
Formula Weight: 130.53
Flash Point: 85°(185°F)
Refractive Index: 1.5310
Melting Point: -2 to 1°
Boiling Point: 173-174°
Beilstein Reference: 110144
Merck Reference: 14,4310
UN No.: 3265
MDL No.: MFCD00003228
Formula:
C₅H₃ClO₂ Boiling Pt: 173…174 °C Melting Pt: –2 to 1 °C Density: 1.325 Flash Pt: 85 °C(185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003228 CAS Number: 527-69-5 EINECS: 208-422-9 UN: 3265 ADR: "8",III Merck Index: 14,4310 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...