2-Amino-4-chloro-6-methylpyrimidine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Chloro-6-methylpyrimidin-2-amine

L06896-06 L06896-14
CAAAAL06896-06EA 54.67 CAD
CAAAAL06896-06 CAAAAL06896-14
2-Amino-4-chloro-6-methylpyrimidine 98%
2-Amino-4-chloro-6-methylpyrimidine

Purity: 98% CAS No.: 5600-21-5 Molecular Formula: C5H6ClN3 Formula Weight: 143.58 Melting Point: 181-186° Beilstein Reference: 114297

Formula: C₅H₆ClN₃
MW: 143.58 g/mol
Melting Pt: 181…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00006091
CAS Number: 5600-21-5
EINECS: 227-018-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR