5-Aminobenzotriazole 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L06660-03
L06660-06
CAAAAL06660-03EA
202.24
CAD
CAAAAL06660-03
CAAAAL06660-06
5-Aminobenzotriazole 96%
5-Aminobenzotriazole
Purity: 96%
CAS No.: 3325-11-9
Molecular Formula: C6H6N4
Formula Weight: 134.14
Odor: Odorless
Melting Point: 157-162°
Beilstein Reference: 121262
MDL No.: MFCD00047209
Formula:
C₆H₆N₄ Melting Pt: 157…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047209 CAS Number: 3325-11-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...