3-Chloro-6-hydrazinopyridazine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L06121-03
L06121-06
CAAAAL06121-03EA
34.44
CAD
CAAAAL06121-03
CAAAAL06121-06
3-Chloro-6-hydrazinopyridazine 98%
3-Chloro-6-hydrazinopyridazine
Purity: 98%
CAS No.: 17284-97-8
Molecular Formula: C4H5ClN4
Formula Weight: 144.56
Melting Point: 134-140°
Beilstein Reference: 125591
Formula:
C₄H₅ClN₄ Melting Pt: 134…140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051740 CAS Number: 17284-97-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...