4-Methoxyphenylhydrazinium chloride 99%

Supplier: Thermo Scientific Chemicals
Warning

L06076-06 L06076-14 L06076-22
CAAAAL06076-06EA 120.55 CAD
CAAAAL06076-06 CAAAAL06076-14 CAAAAL06076-22
4-Methoxyphenylhydrazinium chloride 99%
4-Methoxyphenylhydrazinium chloride

Purity: 99% CAS No.: 19501-58-7 Molecular Formula: C7H10N2O.HCl Formula Weight: 174.63 Melting Point: ca 161° dec. Beilstein Reference: 3566583

Formula: C₇H₁₁ClN₂O
Melting Pt: ca 161 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012945
CAS Number: 19501-58-7
EINECS: 243-115-3
UN: 2811
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR