4-Tritylphenol 98%

Supplier: Thermo Scientific Chemicals
Warning

L06073-06 L06073-14
CAAAAL06073-06EA 71.26 CAD
CAAAAL06073-06 CAAAAL06073-14
4-Tritylphenol 98%
4-Tritylphenol

Purity: 98% CAS No.: 978-86-9 Molecular Formula: C25H20O Formula Weight: 336.43 Melting Point: 276-286° Beilstein Reference: 2290531

Formula: C₂₅H₂₀O
MW: 336.43 g/mol
Melting Pt: 276…286 °C
Storage Temperature: Ambient
MDL Number: MFCD00002364
CAS Number: 978-86-9
EINECS: 213-557-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR