trans,trans-2,4-Dodecadienal 90+% remainder mainly trans, cis isomer
Supplier: Thermo Scientific Chemicals
|
Warning
|
L06002-03
L06002-06
CAAAAL06002-03EA
70.47
CAD
CAAAAL06002-03
CAAAAL06002-06
trans,trans-2,4-Dodecadienal 90+% remainder mainly trans, cis isomer
trans,trans-2,4-Dodecadienal
Purity: 90+%
CAS No.: 21662-16-8
Molecular Formula: C12H20O
Formula Weight: 180.29
Storage Temperature: 2°C to 8°C
Flash Point: 104°(219°F)
Refractive Index: 1.5100
Boiling Point: 130°/6mm
Beilstein Reference: 1708096
MDL No.: MFCD00014678
Formula:
C₁₂H₂₀O Boiling Pt: 130 °C/6 mm Density: 0,863 Flash Pt: 104 °C(219 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00014678 CAS Number: 21662-16-8 EINECS: 244-517-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...