4-(2-Chloroethyl)morpholine hydrochloride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N-(2-Chloroethyl)morpholine hydrochloride, N-(2-Chloroethyl)-Morpholinium Chloride
L05771-22
L05771-36
CAAAAL05771-22EA
59.41
CAD
CAAAAL05771-22
CAAAAL05771-36
4-(2-Chloroethyl)morpholine hydrochloride 99%
4-(2-Chloroethyl)morpholine hydrochloride
Purity: 99%
CAS No.: 3647-69-6
Molecular Formula: C6H12ClNO.HCl
Formula Weight: 186.08
Melting Point: 184-186°
Beilstein Reference: 3684083
UN No.: 2923
MDL No.: MFCD00012797
Formula:
C₆H₁₃Cl₂NO MW: 186.08 g/mol Melting Pt: 184…186 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012797 CAS Number: 3647-69-6 EINECS: 222-881-2 UN: 2923 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...