Chloro(methyl)(diphenyl)silane 97%

Supplier: Thermo Scientific Chemicals
Danger

L04162-09 L04162-18
CAAAAL04162-09EA 72.52 CAD
CAAAAL04162-09 CAAAAL04162-18
Chloro(methyl)(diphenyl)silane 97%
Chloro(methyl)(diphenyl)silane

Fieser: 10,91 12,321 13,74


Purity: 97% CAS No.: 144-79-6 Molecular Formula: C13H13ClSi Formula Weight: 232.79 Flash Point: 141°(285°F) Refractive Index: 1.5740 Odor: Irritating Boiling Point: 294-295° Beilstein Reference: 2937445 UN No.: 2987 MDL No.: MFCD00000498

Formula: C₁₃H₁₃ClSi
MW: 232.78 g/mol
Boiling Pt: 294…295 °C
Density: 1.104
Flash Pt: 141 °C (285 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000498
CAS Number: 144-79-6
EINECS: 205-639-0
UN: 2987
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR