trans,trans-Muconic acid (trans,trans-1,3-butadiene-1,4-dicarboxylic acid) 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L03987-03
L03987-06
CAAAAL03987-03EA
69.68
CAD
CAAAAL03987-03
CAAAAL03987-06
trans,trans-Muconic acid (trans,trans-1,3-butadiene-1,4-dicarboxylic acid) 98+%
trans,trans-Muconic acid (trans,trans-1,3-Butadiene-1,4-dicarboxylic acid)
Purity: 98+%
CAS No.: 3588-17-8
Molecular Formula: C6H6O4
Formula Weight: 142.11
Melting Point: ca 300° dec.
Beilstein Reference: 1722248
Merck Reference: 14,6299
MDL No.: MFCD00002702
Formula:
C₆H₆O₄ Melting Pt: ca 300 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00002702 CAS Number: 3588-17-8 EINECS: 222-724-8 Merck Index: 14,6299 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...