Chloro(diphenyl)silane tech. 90%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
L03602-06
L03602-14
CAAAAL03602-06EA
146.62
CAD
CAAAAL03602-06
CAAAAL03602-14
Chloro(diphenyl)silane tech. 90%, Technical Grade
Chloro(diphenyl)silane
Purity: 90%
CAS No.: 1631-83-0
Molecular Formula: C12H11ClSi
Formula Weight: 218.76
Flash Point: 105°(221°F)
Refractive Index: 1.5790
Boiling Point: 280°
Beilstein Reference: 2937164
UN No.: 2987
MDL No.: MFCD00045144
Formula:
C₁₂H₁₁ClSi MW: 218.76 g/mol Boiling Pt: 280 °C Density: 1.123 Flash Pt: 105 °C (221 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00045144 CAS Number: 1631-83-0 EINECS: 216-634-8 UN: 2987 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...