3,4-Dichlorobenzyl mercaptan 97%

Supplier: Thermo Scientific Chemicals
Danger

L03226-03 L03226-06
CAAAAL03226-03EA 63.99 CAD
CAAAAL03226-03 CAAAAL03226-06
3,4-Dichlorobenzyl mercaptan 97%
3,4-Dichlorobenzyl mercaptan

Purity: 97% CAS No.: 36480-40-7 Molecular Formula: C7H6Cl2S Formula Weight: 193.10 Flash Point: 110°(230°F) Refractive Index: 1.6070 Boiling Point: 152-153°/12mm Beilstein Reference: 3237473 UN No.: 2810 MDL No.: MFCD00039651

Formula: C₇H₆Cl₂S
Boiling Pt: 152…153 °C/12 mm
Density: 1.362
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039651
CAS Number: 36480-40-7
EINECS: 253-054-4
UN: 2810
ADR: "6.1",III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR