2,5-Dibromo-3-methylthiophene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L02693-06
L02693-14
CAAAAL02693-06EA
63.2
CAD
CAAAAL02693-06
CAAAAL02693-14
CAAAAL02693-22
2,5-Dibromo-3-methylthiophene 98%
2,5-Dibromo-3-methylthiophene
Purity: 98%
CAS No.: 13191-36-1
Molecular Formula: C5H4Br2S
Formula Weight: 255.97
Refractive Index: 1.6130
Boiling Point: 226-230°
MDL No.: MFCD00015470
Formula:
C₅H₄Br₂S Boiling Pt: 226…230 °C Density: 1,972 Storage Temperature: Ambient |
MDL Number:
MFCD00015470 CAS Number: 13191-36-1 EINECS: 236-147-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...